Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCTCCTAGGACCCTCCCTCCA[C/T]GAGCCAAGAACATCTGACCCTGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603540 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABBR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABBR1 - gamma-aminobutyric acid type B receptor subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319053.1 | Intron | NP_001305982.1 | ||||
NM_001470.3 | Intron | NP_001461.1 | ||||
NM_021903.2 | Intron | NP_068703.1 | ||||
NM_021904.3 | Intron | NP_068704.2 | ||||
XM_005248982.2 | Intron | XP_005249039.1 | ||||
XM_006715047.3 | Intron | XP_006715110.1 | ||||
XM_011514453.2 | Intron | XP_011512755.1 | ||||
XM_011514455.1 | Intron | XP_011512757.1 | ||||
XM_017010676.1 | Intron | XP_016866165.1 |
Set Membership: |
HapMap |