Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTAAATGCATTATTTGTTATTA[C/T]GACATTATTGTGAAATGATGTAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607307 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FILIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FILIP1 - filamin A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289987.1 | Intron | NP_001276916.1 | ||||
NM_001300866.1 | Intron | NP_001287795.1 | ||||
NM_015687.3 | Intron | NP_056502.1 | ||||
XM_005248713.3 | Intron | XP_005248770.1 | ||||
XM_005248715.4 | Intron | XP_005248772.1 | ||||
XM_011535756.2 | Intron | XP_011534058.1 |
LOC100506804 - uncharacterized LOC100506804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |