Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACAAATGTCTCAGAAATACTT[G/T]ATAATGCTAAATAAGAGTTTATCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607307 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FILIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FILIP1 - filamin A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289987.1 | Intron | NP_001276916.1 | ||||
NM_001300866.1 | Intron | NP_001287795.1 | ||||
NM_015687.3 | Intron | NP_056502.1 | ||||
XM_005248713.3 | Intron | XP_005248770.1 | ||||
XM_005248715.4 | Intron | XP_005248772.1 | ||||
XM_011535756.2 | Intron | XP_011534058.1 |
LOC100506804 - uncharacterized LOC100506804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |