Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCACTGTGCCCTGAGGGAGGG[A/G]GACATGGAGTATGCATGCTTCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRK - fyn related Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002031.2 | Intron | NP_002022.1 | ||||
XM_005266880.4 | Intron | XP_005266937.1 | ||||
XM_005266881.2 | Intron | XP_005266938.1 | ||||
XM_005266882.4 | Intron | XP_005266939.1 | ||||
XM_011535653.2 | Intron | XP_011533955.1 | ||||
XM_011535654.2 | Intron | XP_011533956.1 | ||||
XM_011535655.2 | Intron | XP_011533957.1 | ||||
XM_011535656.2 | Intron | XP_011533958.1 | ||||
XM_017010645.1 | Intron | XP_016866134.1 |
NT5DC1 - 5'-nucleotidase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152729.2 | Intron | NP_689942.2 | ||||
XM_006715378.3 | Intron | XP_006715441.1 |