Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603392 MIM: 614479 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BIK - BCL2 interacting killer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCAT - malonyl-CoA-acyl carrier protein transacylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014507.3 | 2031 | UTR 3 | NP_055322.1 | |||
NM_173467.4 | 2031 | UTR 3 | NP_775738.3 | |||
XM_011530156.2 | 2031 | Intron | XP_011528458.1 |
Set Membership: |
HapMap |