Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197327.1 | Intron | NP_001184256.1 | ||||
NM_001318322.1 | Intron | NP_001305251.1 | ||||
NM_052970.4 | Intron | NP_443202.3 | ||||
XM_011529151.2 | Intron | XP_011527453.1 | ||||
XM_017027632.1 | Intron | XP_016883121.1 | ||||
XM_017027633.1 | Intron | XP_016883122.1 | ||||
XM_017027634.1 | Intron | XP_016883123.1 | ||||
XM_017027635.1 | Intron | XP_016883124.1 |