Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609231 | ||||||||||||||||||||||||||||||||
Literature Links: |
NUDT7 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
NUDT7 - nudix hydrolase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105663.2 | Intron | NP_001099133.1 | ||||
NM_001243657.1 | Intron | NP_001230586.1 | ||||
NM_001243660.1 | Intron | NP_001230589.1 | ||||
NM_001243661.1 | Intron | NP_001230590.1 | ||||
XM_011523032.2 | Intron | XP_011521334.1 | ||||
XM_011523033.2 | Intron | XP_011521335.1 | ||||
XM_017023170.1 | Intron | XP_016878659.1 |
Set Membership: |
HapMap |