Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC130 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC130 - coiled-coil domain containing 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320561.1 | Intron | NP_001307490.1 | ||||
NM_001320564.1 | Intron | NP_001307493.1 | ||||
NM_001320565.1 | Intron | NP_001307494.1 | ||||
NM_001320566.1 | Intron | NP_001307495.1 | ||||
NM_001320567.1 | Intron | NP_001307496.1 | ||||
NM_001320568.1 | Intron | NP_001307497.1 | ||||
NM_001320569.1 | Intron | NP_001307498.1 | ||||
NM_030818.3 | Intron | NP_110445.1 | ||||
XM_005260086.4 | Intron | XP_005260143.1 | ||||
XM_011528326.1 | Intron | XP_011526628.1 |
Set Membership: |
HapMap |