Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAAAACCTACCTGCAGTTGTA[C/T]GAGGCTGTTACTGGAGTAGCAGATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609389 MIM: 610909 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5F PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INPP5F - inositol polyphosphate-5-phosphatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243194.1 | 3142 | Intron | NP_001230123.1 | |||
NM_001243195.1 | 3142 | Intron | NP_001230124.1 | |||
NM_014937.3 | 3142 | Intron | NP_055752.1 |
MCMBP - minichromosome maintenance complex binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256378.1 | 3142 | UTR 3 | NP_001243307.1 | |||
NM_001256379.1 | 3142 | UTR 3 | NP_001243308.1 | |||
NM_024834.3 | 3142 | UTR 3 | NP_079110.1 | |||
XM_005270157.4 | 3142 | UTR 3 | XP_005270214.1 | |||
XM_011540170.1 | 3142 | UTR 3 | XP_011538472.1 | |||
XM_017016663.1 | 3142 | UTR 3 | XP_016872152.1 | |||
XM_017016664.1 | 3142 | UTR 3 | XP_016872153.1 |
Set Membership: |
HapMap |