Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGACCTGAACATTTAAAGTGCT[A/G]TTGGCTAGGTTTCCTTTTCTTCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185861 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SV2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SV2B - synaptic vesicle glycoprotein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167580.1 | Intron | NP_001161052.1 | ||||
NM_001323031.1 | Intron | NP_001309960.1 | ||||
NM_001323032.1 | Intron | NP_001309961.1 | ||||
NM_001323033.1 | Intron | NP_001309962.1 | ||||
NM_001323034.1 | Intron | NP_001309963.1 | ||||
NM_001323036.1 | Intron | NP_001309965.1 | ||||
NM_001323037.1 | Intron | NP_001309966.1 | ||||
NM_001323038.1 | Intron | NP_001309967.1 | ||||
NM_001323039.1 | Intron | NP_001309968.1 | ||||
NM_001323040.1 | Intron | NP_001309969.1 | ||||
NM_014848.5 | Intron | NP_055663.1 | ||||
XM_005254998.3 | Intron | XP_005255055.1 | ||||
XM_017022761.1 | Intron | XP_016878250.1 | ||||
XM_017022762.1 | Intron | XP_016878251.1 |