Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGTCCTCATCCCCAACATAAGC[C/T]TCAGGCTCTCTCCCATCTTCAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607226 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC11 - histone deacetylase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136041.2 | Intron | NP_001129513.1 | ||||
NM_024827.3 | Intron | NP_079103.2 | ||||
XM_005265491.1 | Intron | XP_005265548.1 | ||||
XM_005265492.1 | Intron | XP_005265549.1 | ||||
XM_006713338.1 | Intron | XP_006713401.1 | ||||
XM_011534131.1 | Intron | XP_011532433.1 | ||||
XM_011534132.2 | Intron | XP_011532434.2 | ||||
XM_011534135.1 | Intron | XP_011532437.1 | ||||
XM_017007234.1 | Intron | XP_016862723.1 |
HDAC11-AS1 - HDAC11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |