Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATGGCCGGCCCCTAGCCGGTCC[C/T]GAGGCCTCGCGAGAGCTCCCGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615229 MIM: 603872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1QL4 - complement C1q like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927267 - uncharacterized LOC101927267 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TROAP - trophinin associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100620.1 | Intron | NP_001094090.1 | ||||
NM_001278324.1 | Intron | NP_001265253.1 | ||||
NM_005480.3 | Intron | NP_005471.3 | ||||
XM_006719181.1 | Intron | XP_006719244.1 | ||||
XM_011537723.1 | Intron | XP_011536025.1 | ||||
XM_011537724.1 | Intron | XP_011536026.1 | ||||
XM_011537725.2 | Intron | XP_011536027.1 |