Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602925 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3CB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3CB - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256045.1 | Intron | NP_001242974.1 | ||||
NM_006219.2 | Intron | NP_006210.1 | ||||
XM_005247530.2 | Intron | XP_005247587.1 | ||||
XM_006713659.3 | Intron | XP_006713722.1 | ||||
XM_011512895.2 | Intron | XP_011511197.1 | ||||
XM_011512896.1 | Intron | XP_011511198.1 | ||||
XM_017006619.1 | Intron | XP_016862108.1 | ||||
XM_017006620.1 | Intron | XP_016862109.1 | ||||
XM_017006621.1 | Intron | XP_016862110.1 |
Set Membership: |
HapMap |