Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGAGTCTGCACCCTCAACCGCC[A/G]CTCAACACCACCCCTCATACCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNL4A - thioredoxin like 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303471.2 | Intron | NP_001290400.1 | ||||
NM_001305557.1 | Intron | NP_001292486.1 | ||||
NM_001305563.1 | Intron | NP_001292492.1 | ||||
NM_001305564.1 | Intron | NP_001292493.1 | ||||
NM_006701.4 | Intron | NP_006692.1 |