Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGCCTTCCTTTAACACACTAT[C/T]AATTGTACTCCAAACCCAACAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLGRKT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLGRKT - plasminogen receptor with a C-terminal lysine | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018465.3 | Intron | NP_060935.2 | ||||
XM_005251510.4 | Intron | XP_005251567.1 | ||||
XM_005251512.4 | Intron | XP_005251569.1 | ||||
XM_011517960.2 | Intron | XP_011516262.1 |