Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610166 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQSEC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IQSEC1 - IQ motif and Sec7 domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134382.2 | 5174 | UTR 3 | NP_001127854.1 | |||
NM_014869.6 | 5174 | Intron | NP_055684.3 | |||
XM_011534304.2 | 5174 | UTR 3 | XP_011532606.1 | |||
XM_011534305.2 | 5174 | UTR 3 | XP_011532607.1 | |||
XM_011534306.2 | 5174 | UTR 3 | XP_011532608.1 | |||
XM_011534307.2 | 5174 | UTR 3 | XP_011532609.1 | |||
XM_011534308.2 | 5174 | UTR 3 | XP_011532610.1 | |||
XM_011534310.2 | 5174 | UTR 3 | XP_011532612.1 | |||
XM_011534311.2 | 5174 | UTR 3 | XP_011532613.1 | |||
XM_011534312.2 | 5174 | UTR 3 | XP_011532614.1 | |||
XM_011534313.2 | 5174 | UTR 3 | XP_011532615.1 | |||
XM_011534314.2 | 5174 | UTR 3 | XP_011532616.1 | |||
XM_011534315.2 | 5174 | UTR 3 | XP_011532617.1 | |||
XM_017007581.1 | 5174 | UTR 3 | XP_016863070.1 | |||
XM_017007582.1 | 5174 | UTR 3 | XP_016863071.1 |
Set Membership: |
HapMap |