Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602410 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BRPF1 - bromodomain and PHD finger containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003694.1 | Intron | NP_001003694.1 | ||||
NM_001319049.1 | Intron | NP_001305978.1 | ||||
NM_001319050.1 | Intron | NP_001305979.1 | ||||
NM_004634.2 | Intron | NP_004625.2 | ||||
XM_005265449.1 | Intron | XP_005265506.1 | ||||
XM_005265450.1 | Intron | XP_005265507.1 | ||||
XM_005265452.1 | Intron | XP_005265509.1 | ||||
XM_011534101.1 | Intron | XP_011532403.1 | ||||
XM_011534102.1 | Intron | XP_011532404.1 |
CPNE9 - copine family member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |