Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606525 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SRGAP3 - SLIT-ROBO Rho GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THUMPD3 - THUMP domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114092.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | NP_001107564.1 | |
NM_015453.2 | 794 | Silent Mutation | AGC,AGT | S,S 28 | NP_056268.2 | |
XM_005265021.4 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_005265078.1 | |
XM_005265022.4 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_005265079.1 | |
XM_005265023.3 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_005265080.1 | |
XM_005265024.4 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_005265081.1 | |
XM_006713086.3 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_006713149.1 | |
XM_011533570.2 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_011531872.1 | |
XM_011533571.2 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_011531873.1 | |
XM_017006113.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861602.1 | |
XM_017006114.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861603.1 | |
XM_017006115.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861604.1 | |
XM_017006116.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861605.1 | |
XM_017006117.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861606.1 | |
XM_017006118.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861607.1 | |
XM_017006119.1 | 794 | Silent Mutation | AGC,AGT | S,S 28 | XP_016861608.1 |
Set Membership: |
HapMap Validated |