Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603772 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNC1I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DYNC1I1 - dynein cytoplasmic 1 intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135556.1 | Intron | NP_001129028.1 | ||||
NM_001135557.1 | Intron | NP_001129029.1 | ||||
NM_001278421.1 | Intron | NP_001265350.1 | ||||
NM_001278422.1 | Intron | NP_001265351.1 | ||||
NM_004411.4 | Intron | NP_004402.1 | ||||
XM_011515861.1 | Intron | XP_011514163.1 | ||||
XM_011515862.1 | Intron | XP_011514164.1 | ||||
XM_011515863.2 | Intron | XP_011514165.1 | ||||
XM_017011804.1 | Intron | XP_016867293.1 | ||||
XM_017011805.1 | Intron | XP_016867294.1 | ||||
XM_017011806.1 | Intron | XP_016867295.1 | ||||
XM_017011807.1 | Intron | XP_016867296.1 |
Set Membership: |
HapMap |