Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTGCTTCTAGTGTTTCTAGGG[A/G]TAGATACATGGTTTGGTTACGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605139 MIM: 607743 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCT2 - chaperonin containing TCP1 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198842.1 | Intron | NP_001185771.1 | ||||
NM_006431.2 | Intron | NP_006422.1 |
FRS2 - fibroblast growth factor receptor substrate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-1 - microRNA 3913-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-2 - microRNA 3913-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |