Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610824 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A44 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A44 - solute carrier family 25 member 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286184.1 | Intron | NP_001273113.1 | ||||
NM_014655.3 | Intron | NP_055470.1 | ||||
XM_006711657.3 | Intron | XP_006711720.1 | ||||
XM_011510180.1 | Intron | XP_011508482.1 | ||||
XM_011510181.1 | Intron | XP_011508483.1 | ||||
XM_017002904.1 | Intron | XP_016858393.1 | ||||
XM_017002905.1 | Intron | XP_016858394.1 | ||||
XM_017002906.1 | Intron | XP_016858395.1 | ||||
XM_017002907.1 | Intron | XP_016858396.1 | ||||
XM_017002908.1 | Intron | XP_016858397.1 |
Set Membership: |
HapMap |