Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTCAGACTTTGAATACCAGCT[A/C]TACCACTTTCTTGCTGTGTGCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK10 - potassium two pore domain channel subfamily K member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021161.4 | 4476 | UTR 3 | NP_066984.1 | |||
NM_138317.2 | 4476 | UTR 3 | NP_612190.1 | |||
NM_138318.2 | 4476 | UTR 3 | NP_612191.1 | |||
XM_011536840.2 | 4476 | UTR 3 | XP_011535142.1 |
LOC105370612 - uncharacterized LOC105370612 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |