Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606930 MIM: 607274 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
THOC1 - THO complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005131.2 | Intron | NP_005122.2 | ||||
XM_011525772.2 | Intron | XP_011524074.1 | ||||
XM_011525773.1 | Intron | XP_011524075.1 | ||||
XM_011525774.2 | Intron | XP_011524076.1 | ||||
XM_017026103.1 | Intron | XP_016881592.1 | ||||
XM_017026104.1 | Intron | XP_016881593.1 |
USP14 - ubiquitin specific peptidase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |