Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608770 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLAT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DLAT - dihydrolipoamide S-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001931.4 | 2699 | UTR 3 | NP_001922.2 | |||
XM_011542647.2 | 2699 | UTR 3 | XP_011540949.1 |
PIH1D2 - PIH1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082619.1 | 2699 | Intron | NP_001076088.1 | |||
NM_138789.3 | 2699 | Intron | NP_620144.1 | |||
XM_011542593.2 | 2699 | Intron | XP_011540895.2 | |||
XM_017017201.1 | 2699 | Intron | XP_016872690.1 | |||
XM_017017202.1 | 2699 | Intron | XP_016872691.1 | |||
XM_017017203.1 | 2699 | Intron | XP_016872692.1 | |||
XM_017017204.1 | 2699 | Intron | XP_016872693.1 | |||
XM_017017205.1 | 2699 | Intron | XP_016872694.1 |
Set Membership: |
HapMap |