Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615626 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf41 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C15orf41 - chromosome 15 open reading frame 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130010.2 | Intron | NP_001123482.1 | ||||
NM_001290232.1 | Intron | NP_001277161.1 | ||||
NM_001290233.1 | Intron | NP_001277162.1 | ||||
NM_001321756.1 | Intron | NP_001308685.1 | ||||
NM_001321757.1 | Intron | NP_001308686.1 | ||||
NM_001321758.1 | Intron | NP_001308687.1 | ||||
NM_001321759.1 | Intron | NP_001308688.1 | ||||
NM_001321760.1 | Intron | NP_001308689.1 | ||||
NM_001321761.1 | Intron | NP_001308690.1 | ||||
NM_032499.5 | Intron | NP_115888.1 | ||||
XM_011522111.1 | Intron | XP_011520413.1 | ||||
XM_017022676.1 | Intron | XP_016878165.1 | ||||
XM_017022677.1 | Intron | XP_016878166.1 |
CSNK1A1P1 - casein kinase 1 alpha 1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |