Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCACTCTGCCTGGGGATGGCCT[A/G]TGGTGATTCATGGACTCATGTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616578 MIM: 194541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723809 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102723809 - uncharacterized LOC102723809 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB1 - zinc finger and BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123329.1 | 1056 | Intron | NP_001116801.1 | |||
NM_014950.2 | 1056 | Intron | NP_055765.2 | |||
XM_005267410.1 | 1056 | Intron | XP_005267467.1 | |||
XM_011536566.2 | 1056 | Intron | XP_011534868.1 | |||
XM_011536568.2 | 1056 | Intron | XP_011534870.1 | |||
XM_017021094.1 | 1056 | UTR 5 | XP_016876583.1 | |||
XM_017021095.1 | 1056 | Intron | XP_016876584.1 |
ZBTB25 - zinc finger and BTB domain containing 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |