Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAATTGCAATATCTGCCTTAAG[C/T]GTGGTAACAGAGCATCACAGTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616578 MIM: 194541 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723809 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102723809 - uncharacterized LOC102723809 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB1 - zinc finger and BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123329.1 | Intron | NP_001116801.1 | ||||
NM_014950.2 | Intron | NP_055765.2 | ||||
XM_005267410.1 | Intron | XP_005267467.1 | ||||
XM_011536566.2 | Intron | XP_011534868.1 | ||||
XM_011536568.2 | Intron | XP_011534870.1 | ||||
XM_017021094.1 | Intron | XP_016876583.1 | ||||
XM_017021095.1 | Intron | XP_016876584.1 |
ZBTB25 - zinc finger and BTB domain containing 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |