Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGACCCTGAAAGTTCTGAGGTTT[C/T]AGTCCTCTTCCACCTGGGGTAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R36 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1R36 - protein phosphatase 1 regulatory subunit 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172365.2 | Intron | NP_758953.1 | ||||
XM_005267354.3 | Intron | XP_005267411.1 | ||||
XM_005267355.3 | Intron | XP_005267412.1 | ||||
XM_011536468.2 | Intron | XP_011534770.1 |