Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605956 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOD2 - nucleotide binding oligomerization domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293557.1 | Intron | NP_001280486.1 | ||||
NM_022162.2 | Intron | NP_071445.1 | ||||
XM_005256084.3 | Intron | XP_005256141.1 | ||||
XM_006721242.3 | Intron | XP_006721305.1 | ||||
XM_006721243.3 | Intron | XP_006721306.1 | ||||
XM_011523259.2 | Intron | XP_011521561.1 | ||||
XM_011523260.2 | Intron | XP_011521562.1 | ||||
XM_011523261.1 | Intron | XP_011521563.1 | ||||
XM_017023535.1 | Intron | XP_016879024.1 | ||||
XM_017023536.1 | Intron | XP_016879025.1 | ||||
XM_017023537.1 | Intron | XP_016879026.1 | ||||
XM_017023538.1 | Intron | XP_016879027.1 |
Set Membership: |
HapMap |