Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGGTTTGGGGATATTTCACAG[G/T]GCTGATGGAGGGATAAATTAATCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613346 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHISA9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SHISA9 - shisa family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145204.2 | Intron | NP_001138676.2 | ||||
NM_001145205.1 | Intron | NP_001138677.2 | ||||
XM_005255539.3 | Intron | XP_005255596.2 | ||||
XM_011522642.2 | Intron | XP_011520944.1 |