Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601530 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C5orf45 - chromosome 5 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017987.2 | 2753 | Intron | NP_001017987.1 | |||
NM_016175.3 | 2753 | Intron | NP_057259.2 |
SQSTM1 - sequestosome 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142298.1 | 2753 | UTR 3 | NP_001135770.1 | |||
NM_001142299.1 | 2753 | UTR 3 | NP_001135771.1 | |||
NM_003900.4 | 2753 | UTR 3 | NP_003891.1 | |||
XM_011534683.2 | 2753 | Intron | XP_011532985.1 | |||
XM_017010010.1 | 2753 | Intron | XP_016865499.1 |