Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156565 MIM: 600928 MIM: 614403 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPG - N-methylpurine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015052.2 | 250 | Intron | NP_001015052.1 | |||
NM_001015054.2 | 250 | Intron | NP_001015054.1 | |||
NM_002434.3 | 250 | UTR 5 | NP_002425.2 |
NPRL3 - NPR3 like, GATOR1 complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDF1 - rhomboid 5 homolog 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |