Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600210 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RUNX3 - runt related transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031680.2 | 2595 | UTR 3 | NP_001026850.1 | |||
NM_001320672.1 | 2595 | UTR 3 | NP_001307601.1 | |||
NM_004350.2 | 2595 | UTR 3 | NP_004341.1 | |||
XM_005246024.4 | 2595 | UTR 3 | XP_005246081.1 | |||
XM_011542351.1 | 2595 | UTR 3 | XP_011540653.1 | |||
XM_017002670.1 | 2595 | Intron | XP_016858159.1 |
Set Membership: |
HapMap JSNP |