Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTTGTCTTTGTTTTGTTTATGG[C/T]TAGACTTACAGACTTGGAAAATGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611176 | ||||||||||||||||||||
Literature Links: |
CCDC175 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC175 - coiled-coil domain containing 175 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164399.1 | 3670 | Intron | NP_001157871.1 | |||
XM_011537120.2 | 3670 | UTR 3 | XP_011535422.1 | |||
XM_011537122.2 | 3670 | UTR 3 | XP_011535424.1 | |||
XM_011537124.2 | 3670 | UTR 3 | XP_011535426.1 | |||
XM_011537125.2 | 3670 | Intron | XP_011535427.1 | |||
XM_011537130.2 | 3670 | Intron | XP_011535432.1 | |||
XM_017021621.1 | 3670 | Intron | XP_016877110.1 |
JKAMP - JNK1/MAPK8-associated membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098625.2 | 3670 | UTR 3 | NP_001092095.1 | |||
NM_001284201.1 | 3670 | UTR 3 | NP_001271130.1 | |||
NM_001284202.1 | 3670 | UTR 3 | NP_001271131.1 | |||
NM_001284203.1 | 3670 | UTR 3 | NP_001271132.1 | |||
NM_001284204.1 | 3670 | UTR 3 | NP_001271133.1 | |||
NM_016475.4 | 3670 | UTR 3 | NP_057559.2 | |||
XM_006720171.3 | 3670 | UTR 3 | XP_006720234.1 | |||
XM_017021366.1 | 3670 | UTR 3 | XP_016876855.1 |