Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602382 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLD1 - phospholipase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130081.2 | Intron | NP_001123553.1 | ||||
NM_002662.4 | Intron | NP_002653.1 | ||||
XM_005247533.2 | Intron | XP_005247590.1 | ||||
XM_005247534.2 | Intron | XP_005247591.1 | ||||
XM_011512897.1 | Intron | XP_011511199.1 | ||||
XM_011512898.1 | Intron | XP_011511200.1 | ||||
XM_011512899.2 | Intron | XP_011511201.1 | ||||
XM_017006623.1 | Intron | XP_016862112.1 | ||||
XM_017006624.1 | Intron | XP_016862113.1 |
Set Membership: |
Validated |