Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTTTGTGCTGCTTCAAAACATG[A/G]CAGAGAAGGTAAAAAAGGAAGTGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120950 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8A - complement component 8 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000562.2 | Intron | NP_000553.1 | ||||
XM_011542079.2 | Intron | XP_011540381.1 | ||||
XM_017002234.1 | Intron | XP_016857723.1 |