Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACCTGTGCGCGCTGCAGAACTC[C/G]GTGCCGCTGCCCGCCGTGCGCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP78 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP78 - centrosomal protein 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098802.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | NP_001092272.1 | |
NM_032171.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | NP_115547.1 | |
XM_005252263.4 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_005252320.1 | |
XM_005252265.2 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_005252322.1 | |
XM_005252266.4 | 373 | UTR 5 | XP_005252323.1 | |||
XM_017015192.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_016870681.1 | |
XM_017015193.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_016870682.1 | |
XM_017015194.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_016870683.1 | |
XM_017015195.1 | 373 | Silent Mutation | TCC,TCG | S,S 28 | XP_016870684.1 | |
XM_017015196.1 | 373 | UTR 5 | XP_016870685.1 |