Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614513 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOR1AIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TOR1AIP2 - torsin 1A interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199260.1 | 3102 | UTR 3 | NP_001186189.1 | |||
NM_022347.3 | 3102 | Intron | NP_071742.1 | |||
NM_145034.4 | 3102 | UTR 3 | NP_659471.1 | |||
XM_005244939.4 | 3102 | UTR 3 | XP_005244996.1 | |||
XM_017000474.1 | 3102 | UTR 3 | XP_016855963.1 | |||
XM_017000475.1 | 3102 | UTR 3 | XP_016855964.1 | |||
XM_017000476.1 | 3102 | UTR 3 | XP_016855965.1 | |||
XM_017000477.1 | 3102 | Intron | XP_016855966.1 | |||
XM_017000478.1 | 3102 | Intron | XP_016855967.1 |