Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCATCCGGGCAAAGTTGTAGTA[C/T]CGCCACATGTGAAAGATATTGTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606979 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COG8 - component of oligomeric golgi complex 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIP7 - NIP7, nucleolar pre-rRNA processing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199434.1 | 590 | Intron | NP_001186363.1 | |||
NM_016101.4 | 590 | Intron | NP_057185.1 |
TMED6 - transmembrane p24 trafficking protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144676.3 | 590 | Silent Mutation | CGA,CGG | R,R 178 | NP_653277.2 |
Set Membership: |
HapMap |