Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611405 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RCL1 - RNA terminal phosphate cyclase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286699.1 | Intron | NP_001273628.1 | ||||
NM_001286700.1 | Intron | NP_001273629.1 | ||||
NM_001286701.1 | Intron | NP_001273630.1 | ||||
NM_005772.4 | Intron | NP_005763.3 | ||||
XM_006716715.3 | Intron | XP_006716778.1 | ||||
XM_011517673.2 | Intron | XP_011515975.1 | ||||
XM_017014176.1 | Intron | XP_016869665.1 |
Set Membership: |
HapMap |