Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604028 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC22C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEC22C - SEC22 homolog C, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201572.1 | 4548 | Intron | NP_001188501.1 | |||
NM_001201584.1 | 4548 | Intron | NP_001188513.1 | |||
NM_004206.3 | 4548 | Intron | NP_004197.1 | |||
NM_032970.3 | 4548 | UTR 3 | NP_116752.1 | |||
XM_005265533.1 | 4548 | UTR 3 | XP_005265590.1 | |||
XM_005265534.1 | 4548 | UTR 3 | XP_005265591.1 | |||
XM_005265535.1 | 4548 | UTR 3 | XP_005265592.1 | |||
XM_011534212.1 | 4548 | UTR 3 | XP_011532514.1 | |||
XM_017007438.1 | 4548 | Intron | XP_016862927.1 | |||
XM_017007439.1 | 4548 | Intron | XP_016862928.1 |