Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTTTATATATGAACTAAAAGA[A/C]ATGAACTAAAATTTTCAGTATTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602031 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC112 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC112 - coiled-coil domain containing 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040440.2 | Intron | NP_001035530.1 | ||||
NM_152549.2 | Intron | NP_689762.2 | ||||
XM_005271908.1 | Intron | XP_005271965.1 |
PGGT1B - protein geranylgeranyltransferase type I subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |