Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LONRF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LONRF3 - LON peptidase N-terminal domain and ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031855.2 | 209 | Silent Mutation | GCC,GCT | A,A 14 | NP_001027026.1 | |
NM_001289109.1 | 209 | Intron | NP_001276038.1 | |||
NM_024778.5 | 209 | Silent Mutation | GCC,GCT | A,A 14 | NP_079054.3 | |
XM_005262476.2 | 209 | Silent Mutation | GCC,GCT | A,A 14 | XP_005262533.1 | |
XM_011531398.2 | 209 | Silent Mutation | GCC,GCT | A,A 14 | XP_011529700.1 | |
XM_017029845.1 | 209 | Silent Mutation | GCC,GCT | A,A 14 | XP_016885334.1 |
Set Membership: |
HapMap |