Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610228 MIM: 614241 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPN13 - calpain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144575.2 | 2400 | Intron | NP_653176.2 | |||
XM_011533159.2 | 2400 | Intron | XP_011531461.1 | |||
XM_011533160.2 | 2400 | Intron | XP_011531462.1 | |||
XM_011533161.2 | 2400 | Intron | XP_011531463.1 | |||
XM_011533163.2 | 2400 | Intron | XP_011531465.1 | |||
XM_017005264.1 | 2400 | Intron | XP_016860753.1 | |||
XM_017005265.1 | 2400 | UTR 3 | XP_016860754.1 | |||
XM_017005266.1 | 2400 | Intron | XP_016860755.1 |
LCLAT1 - lysocardiolipin acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002257.2 | 2400 | Intron | NP_001002257.1 | |||
NM_001304445.1 | 2400 | Intron | NP_001291374.1 | |||
NM_001304446.1 | 2400 | Intron | NP_001291375.1 | |||
NM_182551.4 | 2400 | Intron | NP_872357.2 | |||
XM_005264244.1 | 2400 | Intron | XP_005264301.1 | |||
XM_005264245.3 | 2400 | Intron | XP_005264302.1 | |||
XM_011532741.2 | 2400 | Intron | XP_011531043.1 | |||
XM_011532742.2 | 2400 | Intron | XP_011531044.1 | |||
XM_011532743.2 | 2400 | Intron | XP_011531045.1 | |||
XM_011532744.2 | 2400 | Intron | XP_011531046.1 | |||
XM_011532745.1 | 2400 | Intron | XP_011531047.1 | |||
XM_017003746.1 | 2400 | Intron | XP_016859235.1 | |||
XM_017003747.1 | 2400 | Intron | XP_016859236.1 |
Set Membership: |
Validated |