Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF638 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF638 - zinc finger protein 638 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014972.2 | Intron | NP_001014972.1 | ||||
NM_001252612.1 | Intron | NP_001239541.1 | ||||
NM_001252613.1 | Intron | NP_001239542.1 | ||||
NM_014497.4 | Intron | NP_055312.2 | ||||
XM_005264263.1 | Intron | XP_005264320.1 | ||||
XM_006711989.1 | Intron | XP_006712052.1 | ||||
XM_011532768.2 | Intron | XP_011531070.2 | ||||
XM_017003809.1 | Intron | XP_016859298.1 | ||||
XM_017003810.1 | Intron | XP_016859299.1 | ||||
XM_017003811.1 | Intron | XP_016859300.1 | ||||
XM_017003812.1 | Intron | XP_016859301.1 |