Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604531 | ||||||||||||||||||||||||||||||||
Literature Links: |
NCR2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
NCR2 - natural cytotoxicity triggering receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199509.1 | 65 | UTR 5 | NP_001186438.1 | |||
NM_001199510.1 | 65 | UTR 5 | NP_001186439.1 | |||
NM_004828.3 | 65 | UTR 5 | NP_004819.2 | |||
XM_017011500.1 | 65 | Missense Mutation | ACG,ATG | T,M 1 | XP_016866989.1 |