Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300663 MIM: 300880 MIM: 300620 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATG4A - autophagy related 4A cysteine peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD10 - proteasome 26S subunit, non-ATPase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002814.3 | Intron | NP_002805.1 | ||||
NM_170750.2 | Intron | NP_736606.1 |
VSIG1 - V-set and immunoglobulin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |