Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612088 MIM: 606783 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC12A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLEC12A - C-type lectin domain family 12 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207010.1 | Intron | NP_001193939.1 | ||||
NM_001300730.1 | Intron | NP_001287659.1 | ||||
NM_138337.5 | Intron | NP_612210.4 | ||||
NM_201623.3 | Intron | NP_963917.2 | ||||
XM_005253324.2 | Intron | XP_005253381.1 | ||||
XM_006719033.3 | Intron | XP_006719096.1 | ||||
XM_006719035.3 | Intron | XP_006719098.1 | ||||
XM_006719036.3 | Intron | XP_006719099.1 | ||||
XM_011520570.1 | Intron | XP_011518872.1 | ||||
XM_011520571.2 | Intron | XP_011518873.2 | ||||
XM_011520573.2 | Intron | XP_011518875.1 |
CLEC1B - C-type lectin domain family 1 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099431.1 | Intron | NP_001092901.1 | ||||
NM_016509.3 | Intron | NP_057593.3 | ||||
XM_005253382.4 | Intron | XP_005253439.1 | ||||
XM_011520685.2 | Intron | XP_011518987.1 | ||||
XM_011520686.2 | Intron | XP_011518988.1 | ||||
XM_017019395.1 | Intron | XP_016874884.1 | ||||
XM_017019396.1 | Intron | XP_016874885.1 |