Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607646 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB7B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZBTB7B - zinc finger and BTB domain containing 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252406.2 | Intron | NP_001239335.1 | ||||
NM_001256455.1 | Intron | NP_001243384.1 | ||||
XM_006711349.2 | Intron | XP_006711412.2 | ||||
XM_006711353.1 | Intron | XP_006711416.1 | ||||
XM_006711354.1 | Intron | XP_006711417.1 | ||||
XM_006711356.3 | Intron | XP_006711419.1 | ||||
XM_006711357.1 | Intron | XP_006711420.1 | ||||
XM_006711358.1 | Intron | XP_006711421.1 | ||||
XM_006711359.2 | Intron | XP_006711422.1 | ||||
XM_011509598.2 | Intron | XP_011507900.1 | ||||
XM_011509599.2 | Intron | XP_011507901.1 | ||||
XM_017001399.1 | Intron | XP_016856888.1 |