Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACATTGTCCTCTCTCTGGATG[C/T]CTCAGATCTCCCCATCTATTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159558 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLLT3 - MLLT3, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286691.1 | 6296 | UTR 3 | NP_001273620.1 | |||
NM_004529.3 | 6296 | UTR 3 | NP_004520.2 | |||
XM_017014726.1 | 6296 | Intron | XP_016870215.1 | |||
XM_017014727.1 | 6296 | Intron | XP_016870216.1 |
Set Membership: |
HapMap |